CAS RN 689-67-8
CAS RN 689-67-8
IUPAC Name:
(5E)-6,10-dimethylundeca-5,9-dien-2-one
Synonyms:
geranylacetone dihydropseudoionone
Molecular Weight (g/mol):
194.32
Molecular Formula:
C13H22O
InChi Key:
HNZUNIKWNYHEJJ-FMIVXFBMSA-N
SMILES:
CC(C)=CCC\C(C)=C\CCC(C)=O
1
–
1
of
1
results
1
Geranylacetone, (E)+(Z), 97%, (Z)-isomer (nerylacetone) ca 45%
CAS: 689-67-8 Molecular Formula: C13H22O Molecular Weight (g/mol): 194.318 MDL Number: MFCD00008910 InChI Key: HNZUNIKWNYHEJJ-FMIVXFBMSA-N Synonym: geranylacetone,geranyl acetone,e-6,10-dimethylundeca-5,9-dien-2-one,trans-geranylacetone,5e-6,10-dimethylundeca-5,9-dien-2-one,geranylacetone, trans,dihydropseudoionone,6,10-dimethylundeca-5,9-dien-2-one,e-6,10-dimethyl-5,9-undecadien-2-one,unii-9b7ry79u9z PubChem CID: 1549778 ChEBI: CHEBI:67206 IUPAC Name: (5E)-6,10-dimethylundeca-5,9-dien-2-one SMILES: CC(=CCCC(=CCCC(=O)C)C)C