accessibility menu, dialog, popup

UserName

CAS RN 690632-06-5

CH3NNH2

CAS RN 690632-06-5

IUPAC Name: 1-{3-[(4-methylpiperidin-1-yl)methyl]phenyl}methanamine
Synonyms: 1-{3-[(4-methylpiperidin-1-yl)methyl]phenyl}methanamine
Molecular Weight (g/mol): 218.34
Molecular Formula: C14H22N2
InChi Key: ZULQRDYUOOXCFV-UHFFFAOYSA-N
SMILES: CC1CCN(CC2=CC(CN)=CC=C2)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results