CAS RN 6923-52-0
IUPAC Name:
antimony(3+) triacetate
Synonyms:
antimony(III) acetate
Molecular Weight (g/mol):
298.89
Molecular Formula:
C6H9O6Sb
InChi Key:
JVLRYPRBKSMEBF-UHFFFAOYSA-K
SMILES:
[Sb+3].CC([O-])=O.CC([O-])=O.CC([O-])=O
1
–
2
of
2
results
1
Antimony(III) acetate, 97%
CAS: 6923-52-0 Molecular Formula: C6H9O6Sb Molecular Weight (g/mol): 298.892 MDL Number: MFCD00014974 InChI Key: JVLRYPRBKSMEBF-UHFFFAOYSA-K Synonym: antimony triacetate,antimony iii acetate,antimony acetate,acetic acid, antimony 3+ salt,octan antimonity czech,antimony 3+ triacetate,acetic acid, antimony 3+ salt 3:1,acetic acid, trianhydride with antimonic acid h3sbo3,acetic acid, antimony salt,octan antimonity PubChem CID: 23354 IUPAC Name: antimony(3+);triacetate SMILES: CC(=O)[O-].CC(=O)[O-].CC(=O)[O-].[Sb+3]