accessibility menu, dialog, popup

UserName

H3CCH3O

CAS RN 693-54-9

IUPAC Name: decan-2-one
Synonyms: 2-decanone
Molecular Weight (g/mol): 156.27
Molecular Formula: C10H20O
InChi Key: ZAJNGDIORYACQU-UHFFFAOYSA-N
SMILES: CCCCCCCCC(C)=O
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)