accessibility menu, dialog, popup

UserName

CAS RN 6940-78-9

IUPAC Name: 1-bromo-4-chlorobutane
Synonyms: butane, 1-bromo-4-chloro-
Molecular Weight (g/mol): 171.46
Molecular Formula: C4H8BrCl
InChi Key: NIDSRGCVYOEDFW-UHFFFAOYSA-N
SMILES: ClCCCCBr
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)
Boiling Point 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)