accessibility menu, dialog, popup

UserName

CAS RN 69404-97-3

CH3OCH3OSiCH3H3CH3CCH3H3C(E/Z)

CAS RN 69404-97-3

IUPAC Name: 4-[(tert-butyldimethylsilyl)oxy]pent-3-en-2-one
Synonyms: 4-[(tert-butyldimethylsilyl)oxy]pent-3-en-2-one
Molecular Weight (g/mol): 214.38
Molecular Formula: C11H22O2Si
InChi Key: ZRVOCNOYAJIAAP-UHFFFAOYSA-N
SMILES: CC(=O)C=C(C)O[Si](C)(C)C(C)(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results