accessibility menu, dialog, popup

UserName

CAS RN 6954-64-9

IUPAC Name: 3-[(2,4-difluorophenyl)carbamoyl]prop-2-enoic acid
Synonyms: 3-[(2,4-difluorophenyl)carbamoyl]prop-2-enoic acid
Molecular Weight (g/mol): 227.17
Molecular Formula: C10H7F2NO3
InChi Key: DVBSHLGHHLTWPZ-UHFFFAOYSA-N
SMILES: OC(=O)C=CC(=O)NC1=C(F)C=C(F)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results