accessibility menu, dialog, popup

UserName

CAS RN 695816-47-8

CAS RN 695816-47-8

IUPAC Name: 1-[2-(1,3-dioxolan-2-yl)phenyl]-2,5-dimethylphospholane
Synonyms: 1-[2-(1,3-dioxolan-2-yl)phenyl]-2,5-dimethylphospholane
Molecular Weight (g/mol): 264.31
Molecular Formula: C15H21O2P
InChi Key: JAKXTLMNIHPLEU-UHFFFAOYNA-N
SMILES: CC1CCC(C)P1C1=CC=CC=C1C1OCCO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results