accessibility menu, dialog, popup

UserName

CAS RN 696-45-7

IUPAC Name: 6-methoxypyrimidin-4-amine
Synonyms: 6-methoxypyrimidin-4-amine
Molecular Weight (g/mol): 125.13
Molecular Formula: C5H7N3O
InChi Key: VELRBZDRGTVGGT-UHFFFAOYSA-N
SMILES: COC1=CC(N)=NC=N1
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)