accessibility menu, dialog, popup

UserName

CAS RN 6960-42-5

ONOHN

CAS RN 6960-42-5

IUPAC Name: 7-nitro-1H-indole
Synonyms: 7-nitroindole
Molecular Weight (g/mol): 162.15
Molecular Formula: C8H6N2O2
InChi Key: LZJGQIVWUKFTRD-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=C2NC=CC2=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results