accessibility menu, dialog, popup

UserName

CH3H3CCH3OON(S)O

CAS RN 69610-41-9

IUPAC Name: tert-butyl (2S)-2-formylpyrrolidine-1-carboxylate
Synonyms: tert-butyl (2S)-2-formylpyrrolidine-1-carboxylate
Molecular Weight (g/mol): 199.25
Molecular Formula: C10H17NO3
InChi Key: YDBPZCVWPFMBDH-QMMMGPOBSA-N
SMILES: CC(C)(C)OC(=O)N1CCC[C@H]1C=O
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)