accessibility menu, dialog, popup

UserName

CAS RN 6967-29-9

H3CCH3HNOCl

CAS RN 6967-29-9

IUPAC Name: 2-chloro-N-(2,6-diethylphenyl)acetamide
Synonyms: 2-chloro-N-(2,6-diethylphenyl)acetamide
Molecular Weight (g/mol): 225.72
Molecular Formula: C12H16ClNO
InChi Key: LBJVHMAYBNQJBK-UHFFFAOYSA-N
SMILES: CCC1=CC=CC(CC)=C1NC(=O)CCl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results