accessibility menu, dialog, popup

UserName

CH3O(S)HNOSSNH2OOHO(R)SNOHOOSNNNNCH3(?)

CAS RN 69712-56-7

IUPAC Name: (6R,7S)-7-{4-[carbamoyl(carboxy)methylidene]-1,3-dithietane-2-amido}-7-methoxy-3-{[(1-methyl-1H-1,2,3,4-tetrazol-5-yl)sulfanyl]methyl}-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid
Synonyms: cefotetan
Molecular Weight (g/mol): 575.6
Molecular Formula: C17H17N7O8S4
InChi Key: SRZNHPXWXCNNDU-IXOPCIAXSA-N
SMILES: CO[C@]1(NC(=O)C2SC(S2)=C(C(N)=O)C(O)=O)[C@H]2SCC(CSC3=NN=NN3C)=C(N2C1=O)C(O)=O
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)