CAS RN 6975-98-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-Methyldecane, TRC
CAS: 6975-98-0 Molecular Formula: C11H24 Molecular Weight (g/mol): 156.31 Synonym: Isoundecane,Isopah H,Isopar G IUPAC Name: 2-methyldecane SMILES: CCCCCCCCC(C)C