CAS RN 6976-17-6
IUPAC Name:
4-(methylamino)butanoic acid hydrochloridyl
Synonyms:
4-(methylamino)butyric acid hydrochloridyl
Molecular Weight (g/mol):
152.6
Molecular Formula:
C5H11ClNO2
InChi Key:
GYSZPWFLNPONPB-UHFFFAOYSA-N
SMILES:
[Cl].CNCCCC(O)=O
1
–
1
of
1
results
1
4-(Methylamino)butanoic Acid Hydrochloride, TRC
CAS: 6976-17-6 Molecular Formula: C5H11NO2 . HCl Molecular Weight (g/mol): 153.61 Synonym: 4-(Methylamino)butanoic Acid Hydrochloride,4-(Methylamino)butyric Acid Hydrochloride IUPAC Name: 4-(methylamino)butanoic acid;hydrochloride SMILES: Cl.CNCCCC(=O)O