accessibility menu, dialog, popup

UserName

CAS RN 703-56-0

IUPAC Name: 3-phenyl-1,3-oxazolidin-2-one
Synonyms: 3-phenyl-1,3-oxazolidin-2-one
Molecular Weight (g/mol): 163.18
Molecular Formula: C9H9NO2
InChi Key: NCTCGHLIHJJIBK-UHFFFAOYSA-N
SMILES: O=C1OCCN1C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results