CAS RN 706753-75-5
CAS RN 706753-75-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
9,13b-Dehydro Epinastine, TRC
CAS: 706753-75-5 Molecular Formula: C16 H13 N3 Molecular Weight (g/mol): 247.29 Synonym: 9H-Dibenz[c,f]imidazo[1,5-a]azepin-3-amine,9,13b-Dehydro Epinastine,9,13b-Dehydroepinastine SMILES: Nc1ncc2c3ccccc3Cc4ccccc4n12