CAS RN 70685-15-3
CAS RN 70685-15-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
1,4,7-Trioxacyclohexadecane-8,16-dione, TRC
CAS: 70685-15-3 Molecular Formula: C13H22O5 Molecular Weight (g/mol): 258.31 IUPAC Name: 1,4,7-trioxacyclohexadecane-8,16-dione SMILES: O=C1CCCCCCCC(=O)OCCOCCO1