accessibility menu, dialog, popup

UserName

FOFFF

CAS RN 708-64-5

IUPAC Name: 2,2,2-trifluoro-1-(3-fluorophenyl)ethan-1-one
Synonyms: 2,2,2-trifluoro-1-(3-fluorophenyl)ethanone
Molecular Weight (g/mol): 192.11
Molecular Formula: C8H4F4O
InChi Key: NQSPTMFCJGKOQJ-UHFFFAOYSA-N
SMILES: FC1=CC=CC(=C1)C(=O)C(F)(F)F
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results