CAS RN 709-50-2
IUPAC Name:
2-(hydroxymethyl)-6-methoxyoxane-3,4,5-triol
Synonyms:
methyl-α-D-mannoside methyl mannoside mannoside, methyl
Molecular Weight (g/mol):
194.18
Molecular Formula:
C7H14O6
InChi Key:
HOVAGTYPODGVJG-UHFFFAOYNA-N
SMILES:
COC1OC(CO)C(O)C(O)C1O
1
–
1
of
1
results
1
Methyl Beta-D-Glucopyranoside, TRC
CAS: 709-50-2 Molecular Formula: C7 H14 O6 Molecular Weight (g/mol): 194.18 IUPAC Name: (2R,3S,4S,5R,6R)-2-(hydroxymethyl)-6-methoxyoxane-3,4,5-triol SMILES: CO[C@@H]1O[C@H](CO)[C@@H](O)[C@H](O)[C@H]1O