accessibility menu, dialog, popup

UserName

CoCH3OOCH3OO

CAS RN 71-48-7

IUPAC Name: λ²-cobalt(2+) diacetate
Synonyms: cobalt(II) acetate
Molecular Weight (g/mol): 177.02
Molecular Formula: C4H6CoO4
InChi Key: QAHREYKOYSIQPH-UHFFFAOYSA-L
SMILES: [Co++].CC([O-])=O.CC([O-])=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results