accessibility menu, dialog, popup

UserName

CH3SeSeH3C

CAS RN 7101-31-7

IUPAC Name: (methyldiselanyl)methane
Synonyms: dimethyl diselenide
Molecular Weight (g/mol): 188.01
Molecular Formula: C2H6Se2
InChi Key: VLXBWPOEOIIREY-UHFFFAOYSA-N
SMILES: C[Se][Se]C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results