accessibility menu, dialog, popup

UserName

CAS RN 71294-03-6

FOHN

CAS RN 71294-03-6

IUPAC Name: 7-fluoro-2,3-dihydro-1H-indol-2-one
Synonyms: 7-fluoro-1,3-dihydroindol-2-one
Molecular Weight (g/mol): 151.14
Molecular Formula: C8H6FNO
InChi Key: VMUIOEOYZHJLEZ-UHFFFAOYSA-N
SMILES: FC1=CC=CC2=C1NC(=O)C2
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results