accessibility menu, dialog, popup

UserName

CAS RN 7143-82-0

IUPAC Name: 6-methyl-2,3-dihydropyridazin-3-one hydrate
Synonyms: 6-methyl-2H-pyridazin-3-one hydrate
Molecular Weight (g/mol): 128.13
Molecular Formula: C5H8N2O2
InChi Key: DPIVSCHNBCFVEA-UHFFFAOYSA-N
SMILES: O.CC1=NNC(=O)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results