CAS RN 7149-42-0
IUPAC Name:
1-(1-methylpiperidin-4-yl)methanamine
Synonyms:
1-(1-methylpiperidin-4-yl)methanamine
Molecular Weight (g/mol):
128.22
Molecular Formula:
C7H16N2
InChi Key:
AGTPSAZJSOQXHJ-UHFFFAOYSA-N
SMILES:
CN1CCC(CN)CC1
1
–
2
of
2
results
2
(1-Methyl-4-piperidnyl)methanamine, 97%, Thermo Scientific™
CAS: 7149-42-0 Molecular Formula: C7H16N2 Molecular Weight (g/mol): 128.22 MDL Number: MFCD05022430 InChI Key: AGTPSAZJSOQXHJ-UHFFFAOYSA-N Synonym: 1-methylpiperidin-4-yl methanamine,1-methyl-piperidin-4-methylamine,1-methyl-4-piperidinyl methanamine,4-piperidinemethanamine, 1-methyl,1-1-methylpiperidin-4-yl methanamine,1-methyl-4-aminomethyl piperidine,4-aminomethyl-1-methylpiperidine,c-1-methyl-piperidin-4-yl-methylamine,1-methylpiperidine-4-methylamine PubChem CID: 81574 IUPAC Name: (1-methylpiperidin-4-yl)methanamine SMILES: CN1CCC(CN)CC1