accessibility menu, dialog, popup

UserName

INHN

CAS RN 71759-89-2

IUPAC Name: 5-iodo-1H-imidazole
Synonyms: 4-iodo-3H-imidazole
Molecular Weight (g/mol): 193.98
Molecular Formula: C3H3IN2
InChi Key: BHCMXJKPZOPRNN-UHFFFAOYSA-N
SMILES: IC1=CN=CN1
Molecular Weight (g/mol) 
  • (3)
  • (5)
Quantity 
  • (3)
  • (1)
  • (1)
  • (3)
Percent Purity 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results