accessibility menu, dialog, popup

UserName

ONOBr

CAS RN 71897-83-1

IUPAC Name: 4-[(5-bromofuran-2-yl)methyl]morpholine
Synonyms: 4-[(5-bromofuran-2-yl)methyl]morpholine
Molecular Weight (g/mol): 246.1
Molecular Formula: C9H12BrNO2
InChi Key: KNWZHIWPPIXAJQ-UHFFFAOYSA-N
SMILES: BrC1=CC=C(CN2CCOCC2)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results