accessibility menu, dialog, popup

UserName

H3COOSNH2NH3C

CAS RN 7210-76-6

IUPAC Name: ethyl 2-amino-4-methyl-1,3-thiazole-5-carboxylate
Synonyms: ethyl 2-amino-4-methyl-1,3-thiazole-5-carboxylate
Molecular Weight (g/mol): 186.23
Molecular Formula: C7H10N2O2S
InChi Key: WZHUPCREDVWLKC-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=C(C)N=C(N)S1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results