accessibility menu, dialog, popup

UserName

H3COCH3OMn

CAS RN 7245-20-7

IUPAC Name: manganese(2+) dimethanolate
Synonyms: manganese(2+) bis(methoxide)
Molecular Weight (g/mol): 117.01
Molecular Formula: C2H6MnO2
InChi Key: UQIRCVPINUNHQY-UHFFFAOYSA-N
SMILES: [Mn++].C[O-].C[O-]
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results