CAS RN 72521-19-8
CAS RN 72521-19-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
3-Carbamoyl-1-Alpha-D-ribofuranosylpyridinium Chloride, TRC
CAS: 72521-19-8 Molecular Formula: C11H15ClN2O5 Molecular Weight (g/mol): 290.7 Synonym: Pyridinium, 3-(aminocarbonyl)-1-α-D-ribofuranosyl-, chloride (9CI),3-Carbamoyl-1-α-D-ribofuranosylpyridinium chloride (7CI),3-(Aminocarbonyl)-1-α-D-ribofuranosylpyridinium chloride,N-(α-D-Ribosyl)nicotinamide chloride,Nicotinamide α-D-riboside chloride IUPAC Name: 1-[(2S,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)oxolan-2-yl]pyridin-1-ium-3-carboxamide;chloride SMILES: [Cl-].NC(=O)c1ccc[n+](c1)[C@H]2O[C@H](CO)[C@@H](O)[C@H]2O