accessibility menu, dialog, popup

UserName

NOOH

CAS RN 72548-79-9

IUPAC Name: 3-(pyrrolidin-1-yl)benzoic acid
Synonyms: 3-(pyrrolidin-1-yl)benzoic acid
Molecular Weight (g/mol): 191.23
Molecular Formula: C11H13NO2
InChi Key: HRVWGEKZONYEMK-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC(=CC=C1)N1CCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results