accessibility menu, dialog, popup

UserName

ClSO

CAS RN 7283-96-7

IUPAC Name: 5-chlorothiophene-2-carbaldehyde
Synonyms: 5-chlorothiophene-2-carbaldehyde
Molecular Weight (g/mol): 146.59
Molecular Formula: C5H3ClOS
InChi Key: VWYFITBWBRVBSW-UHFFFAOYSA-N
SMILES: ClC1=CC=C(S1)C=O
Boiling Point 
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)