accessibility menu, dialog, popup

UserName

CAS RN 7292-73-1

NH2OHOF

CAS RN 7292-73-1

IUPAC Name: 2-amino-2-(4-fluorophenyl)acetic acid
Synonyms: amino(4-fluorophenyl)acetic acid
Molecular Weight (g/mol): 169.16
Molecular Formula: C8H8FNO2
InChi Key: JKFYKCYQEWQPTM-UHFFFAOYNA-N
SMILES: NC(C(O)=O)C1=CC=C(F)C=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results