accessibility menu, dialog, popup

UserName

CH3H3COClO

CAS RN 7298-89-7

IUPAC Name: 2-chloro-5,5-dimethylcyclohexane-1,3-dione
Synonyms: 2-chloro-5,5-dimethylcyclohexane-1,3-dione
Molecular Weight (g/mol): 174.62
Molecular Formula: C8H11ClO2
InChi Key: VOBIHUAWDXUCPH-UHFFFAOYSA-N
SMILES: CC1(C)CC(=O)C(Cl)C(=O)C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results