accessibility menu, dialog, popup

UserName

ClH3CNH3

CAS RN 7311-30-0

IUPAC Name: tridecan-1-aminium chloride
Synonyms: tridecan-1-aminium chloride
Molecular Weight (g/mol): 235.84
Molecular Formula: C13H30ClN
InChi Key: XDRDMVYFQARLNH-UHFFFAOYSA-N
SMILES: [Cl-].CCCCCCCCCCCCC[NH3+]
Molecular Weight (g/mol) 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)