accessibility menu, dialog, popup

UserName

H3CSiH3CCH3OI

CAS RN 732306-25-1

IUPAC Name: (3-iodophenoxy)trimethylsilane
Synonyms: 3-iodophenoxytrimethylsilane
Molecular Weight (g/mol): 292.19
Molecular Formula: C9H13IOSi
InChi Key: GJDJYHHPFQRAHK-UHFFFAOYSA-N
SMILES: C[Si](C)(C)OC1=CC(I)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results