accessibility menu, dialog, popup

UserName

CAS RN 73397-28-1

H3CCH3OOHOHNHOOH3CH3CCH3

CAS RN 73397-28-1

IUPAC Name: 3-{[(tert-butoxy)carbonyl]amino}-2-hydroxy-5-methylhexanoic acid
Synonyms: 3-[(tert-butoxycarbonyl)amino]-2-hydroxy-5-methylhexanoic acid
Molecular Weight (g/mol): 261.32
Molecular Formula: C12H23NO5
InChi Key: DJZCWTDKDFJARG-UHFFFAOYNA-N
SMILES: CC(C)CC(NC(=O)OC(C)(C)C)C(O)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results