CAS RN 73509-22-5
CAS RN 73509-22-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Indole-4,5,6,7-d4, TRC
CAS: 73509-22-5 Molecular Formula: C8H3D4N Molecular Weight (g/mol): 121.17 Synonym: 1H-Indole-d4, 1-Azaindene-d4,1-Benzazole-d4,2,3-Benzopyrrole-d4,Benzo[b]pyrrole-d4,Ketole-d4,NSC 1964-d4 SMILES: [2H]C1=C2C(NC=C2)=C([2H])C([2H])=C1[2H]