accessibility menu, dialog, popup

UserName

CH2SnH2CH2CH2C

CAS RN 7393-43-3

IUPAC Name: tetrakis(prop-2-en-1-yl)stannane
Synonyms: tetrakis(prop-2-en-1-yl)stannane
Molecular Weight (g/mol): 283
Molecular Formula: C12H20Sn
InChi Key: XJPKDRJZNZMJQM-UHFFFAOYSA-N
SMILES: C=CC[Sn](CC=C)(CC=C)CC=C
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)