CAS RN 73956-15-7
IUPAC Name:
2,2-diethoxyethanethioamide
Synonyms:
2,2-diethoxyethanethioamide
Molecular Weight (g/mol):
163.24
Molecular Formula:
C6H13NO2S
InChi Key:
MQSDGAKLSVITHP-UHFFFAOYSA-N
SMILES:
CCOC(OCC)C(N)=S
1
–
1
of
1
results
1
2,2-Diethoxyethanethioamide, 97%, Thermo Scientific™
CAS: 73956-15-7 Molecular Formula: C6H13NO2S Molecular Weight (g/mol): 163.235 MDL Number: MFCD06658986 InChI Key: MQSDGAKLSVITHP-UHFFFAOYSA-N PubChem CID: 2794734 IUPAC Name: 2,2-diethoxyethanethioamide SMILES: CCOC(C(=S)N)OCC