accessibility menu, dialog, popup

UserName

CH3H3CO(S)OOOO

CAS RN 73991-95-4

IUPAC Name: 2-[(4S)-2,2-dimethyl-5-oxo-1,3-dioxolan-4-yl]acetate
Synonyms: 2-[(4S)-2,2-dimethyl-5-oxo-1,3-dioxolan-4-yl]acetate
Molecular Weight (g/mol): 173.15
Molecular Formula: C7H9O5
InChi Key: IDQOCLIWDMZKBZ-BYPYZUCNSA-M
SMILES: CC1(C)O[C@@H](CC([O-])=O)C(=O)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results