accessibility menu, dialog, popup

UserName

CAS RN 7423-31-6

BrH3CNSCH3SNH3C(Z)(E/Z)

CAS RN 7423-31-6

IUPAC Name: 1-ethyl-2-{3-[(2Z)-1-ethyl-1H,2H-naphtho[1,2-d][1,3]thiazol-2-ylidene]-2-methylprop-1-en-1-yl}naphtho[1,2-d][1,3]thiazol-1-ium bromide
Synonyms: 1-ethyl-2-{3-[(2Z)-1-ethylnaphtho[1,2-d][1,3]thiazol-2-ylidene]-2-methylprop-1-en-1-yl}naphtho[1,2-d][1,3]thiazol-1-ium bromide
Molecular Weight (g/mol): 559.58
Molecular Formula: C30H27BrN2S2
InChi Key: MPBRYMWMMKKRGC-UHFFFAOYSA-M
SMILES: [Br-].CCN1\C(SC2=CC=C3C=CC=CC3=C12)=C\C(C)=CC1=[N+](CC)C2=C(S1)C=CC1=CC=CC=C21
Molecular Weight (g/mol) 
  • (1)
  • (3)
Color 
  • (3)
Grade 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (4)
  • (1)
  • (1)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)