accessibility menu, dialog, popup

UserName

CAS RN 7432-21-5

IUPAC Name: (2S)-2-{[(benzyloxy)carbonyl]amino}-3-(1H-indol-3-yl)propanoate
Synonyms: (2S)-2-{[(benzyloxy)carbonyl]amino}-3-(1H-indol-3-yl)propanoate
Molecular Weight (g/mol): 337.36
Molecular Formula: C19H17N2O4
InChi Key: AHYFYYVVAXRMKB-KRWDZBQOSA-M
SMILES: [O-]C(=O)[C@H](CC1=CNC2=CC=CC=C12)NC(=O)OCC1=CC=CC=C1
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)