accessibility menu, dialog, popup

UserName

CH3ClNH2O

CAS RN 7462-73-9

IUPAC Name: 2-chlorobutanamide
Synonyms: 2-chlorobutanamide
Molecular Weight (g/mol): 121.56
Molecular Formula: C4H8ClNO
InChi Key: VJIOSCMTZVXQQU-UHFFFAOYNA-N
SMILES: CCC(Cl)C(N)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results