accessibility menu, dialog, popup

UserName

CAS RN 74852-81-6

CH3NNH2N

CAS RN 74852-81-6

IUPAC Name: 4-(2-methyl-1H-imidazol-1-yl)aniline
Synonyms: 4-(2-methylimidazol-1-yl)aniline
Molecular Weight (g/mol): 173.22
Molecular Formula: C10H11N3
InChi Key: IEZCMVRWKNEHJB-UHFFFAOYSA-N
SMILES: CC1=NC=CN1C1=CC=C(N)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results