accessibility menu, dialog, popup

UserName

ClCH3NCH3CH3H3C

CAS RN 75-57-0

IUPAC Name: tetramethylazanium chloride
Synonyms: tetramethylammonium chloride
Molecular Weight (g/mol): 109.6
Molecular Formula: C4H12ClN
InChi Key: OKIZCWYLBDKLSU-UHFFFAOYSA-M
SMILES: [Cl-].C[N+](C)(C)C
Molecular Weight (g/mol) 
  • (4)
  • (5)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (3)
  • (4)
Physical Form 
  • (4)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (4)
  • (3)
  • (1)
  • (4)
  • (5)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (4)
  • (4)
  • (1)