accessibility menu, dialog, popup

UserName

CAS RN 7517-76-2

IUPAC Name: 1,4-diisocyanatocyclohexane
Synonyms: 1,4-diisocyanatocyclohexane
Molecular Weight (g/mol): 166.18
Molecular Formula: C8H10N2O2
InChi Key: CDMDQYCEEKCBGR-UHFFFAOYSA-N
SMILES: O=C=NC1CCC(CC1)N=C=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results