accessibility menu, dialog, popup

UserName

CAS RN 75178-87-9

H3CH3CCH3OONHOH

CAS RN 75178-87-9

IUPAC Name: tert-butyl N-(4-hydroxybutyl)carbamate
Synonyms: tert-butyl N-(4-hydroxybutyl)carbamate
Molecular Weight (g/mol): 189.26
Molecular Formula: C9H19NO3
InChi Key: LIYMTLVBAVHPBU-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NCCCCO

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)