accessibility menu, dialog, popup

UserName

CH3NOO

CAS RN 7529-22-8

IUPAC Name: 4-methylmorpholin-4-ium-4-olate
Synonyms: methyl morpholine oxide
Molecular Weight (g/mol): 117.15
Molecular Formula: C5H11NO2
InChi Key: LFTLOKWAGJYHHR-UHFFFAOYSA-N
SMILES: C[N+]1([O-])CCOCC1
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)
  • (3)
  • (6)