accessibility menu, dialog, popup

UserName

CAS RN 7536-55-2

IUPAC Name: (2S)-2-{[(tert-butoxy)carbonyl]amino}-3-carbamoylpropanoic acid
Synonyms: Boc-Asn
Molecular Weight (g/mol): 232.24
Molecular Formula: C9H16N2O5
InChi Key: FYYSQDHBALBGHX-YFKPBYRVSA-N
SMILES: CC(C)(C)OC(=O)N[C@@H](CC(N)=O)C(O)=O
Molecular Weight (g/mol) 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)